ADVH14DBD03E AChemBlock

Br-PEG5-OH

  • MDL number MFCD18252202
  • Cas Number : 957205-14-0
  • Molecular Weight : 301.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/br-peg5-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P34817
Availability details for
Quantity
  • Availability
  • Add to Cart