ADVH14DBD045 AChemBlock

I-PEG4-OH

  • MDL number MFCD30468746
  • Cas Number : 136399-06-9
  • Molecular Weight : 304.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/i-peg4-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41510
Availability details for
Quantity
  • Availability
  • Add to Cart