ADVH14DBD05B AChemBlock

BocNH-PEG4-CH2CH2Br

  • MDL number MFCD31536756
  • Cas Number : 1392499-32-9
  • Molecular Weight : 400.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/bocnh-peg4-ch2ch2br.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41408
Availability details for
Quantity
  • Availability
  • Add to Cart