ADVH14DBD05E AChemBlock

Propargyl-PEG5-OH

  • MDL number MFCD22201537
  • Cas Number : 87450-10-0
  • Molecular Weight : 232.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/propargyl-peg5-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35575
Availability details for
Quantity
  • Availability
  • Add to Cart