ADVH14DBD066 AChemBlock

Heptaethylene glycol

  • MDL number MFCD00002876
  • Cas Number : 5617-32-3
  • Molecular Weight : 326.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/heptaethylene-glycol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P37406
Availability details for
Quantity
  • Availability
  • Add to Cart