ADVH14DBD068 AChemBlock

Nonaethylene glycol

  • MDL number MFCD00698692
  • Cas Number : 3386-18-3
  • Molecular Weight : 414.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/nonaethylene-glycol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P38725
Availability details for
Quantity
  • Availability
  • Add to Cart