ADVH14DBD081 AChemBlock

2-(2-Propyn-1-yloxy)ethanamine

  • MDL number MFCD16713446
  • Cas Number : 122116-12-5
  • Molecular Weight : 99.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-propyn-1-yloxy-ethanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P42183
Availability details for
Quantity
  • Availability
  • Add to Cart