ADVH14DBD091 AChemBlock

Mal-PEG2-CH2CH2COOH

  • MDL number MFCD23726620
  • Cas Number : 1374666-32-6
  • Molecular Weight : 257.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/mal-peg2-ch2ch2cooh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P41477
Availability details for
Quantity
  • Availability
  • Add to Cart