ADVH14DBD0A8 AChemBlock

HOOCCH2O-PEG4-CH2COOH

  • MDL number MFCD29079379
  • Cas Number : 77855-75-5
  • Molecular Weight : 310.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/hoocch2o-peg4-ch2cooh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36094
Availability details for
Quantity
  • Availability
  • Add to Cart