ADVH14DBD0B0 AChemBlock

H2N-PEG5-CH2CH2NH2

  • MDL number MFCD07369626
  • Cas Number : 72236-26-1
  • Molecular Weight : 280.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/h2n-peg5-ch2ch2nh2.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P36419
Availability details for
Quantity
  • Availability
  • Add to Cart