ADVH14DBD0B9 AChemBlock

Propargyl-PEG2-NHBoc

  • MDL number MFCD22418469
  • Cas Number : 869310-84-9
  • Molecular Weight : 243.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/propargyl-peg2-nhboc.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P35632
Availability details for
Quantity
  • Availability
  • Add to Cart