ADVH14DBD0BA AChemBlock

3,3'-Oxybis-1-propanol

  • MDL number MFCD00051023
  • Cas Number : 2396-61-4
  • Molecular Weight : 134.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-3-oxybis-1-propanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P39459
Availability details for
Quantity
  • Availability
  • Add to Cart