ADVH14DBD0BF AChemBlock

3-chloro-6-nitro-1H-indazole

  • MDL number MFCD00010741
  • Cas Number : 50593-68-5
  • Molecular Weight : 197.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-chloro-6-nitro-1h-indazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43259
Availability details for
Quantity
  • Availability
  • Add to Cart