ADVH14DBD0EA AChemBlock

1,3-bis(2-chloroethyl)urea

  • MDL number MFCD00068573
  • Cas Number : 2214-72-4
  • Molecular Weight : 185.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-bis-2-chloroethyl-urea.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43328
Availability details for
Quantity
  • Availability
  • Add to Cart