ADVH14DBD14B AChemBlock

2,4-dimethyloxazole-5-carboxylic acid

  • MDL number MFCD06011090
  • Cas Number : 2510-37-4
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-4-dimethyloxazole-5-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43319
Availability details for
Quantity
  • Availability
  • Add to Cart