ADVH14DBD164 AChemBlock

6,7-dihydroquinolin-8(5H)-one

  • MDL number MFCD03839916
  • Cas Number : 56826-69-8
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-7-dihydroquinolin-8-5h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43242
Availability details for
Quantity
  • Availability
  • Add to Cart