ADVH14DBD168 AChemBlock

3-Ethoxy-2-fluorobenzaldehyde

  • MDL number MFCD24678362
  • Cas Number : 1204176-29-3
  • Molecular Weight : 168.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-ethoxy-2-fluorobenzaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43506
Availability details for
Quantity
  • Availability
  • Add to Cart