ADVH14DBD1D3 AChemBlock

(S)-4-(1-carboxy-2-(4-hydroxyphenyl)ethylamino)-4-oxobutanoic acid

  • MDL number MFCD18252486
  • Cas Number : 374816-32-7
  • Molecular Weight : 281.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-4-1-carboxy-2-4-hydroxyphenyl-ethylamino-4-oxobutanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43283
Availability details for
Quantity
  • Availability
  • Add to Cart