ADVH14DBD1D5 AChemBlock

(1r,4r)-4-(methoxycarbonyl)bicyclo[2.1.1]hexane-1-carboxylic acid

  • MDL number MFCD30174613
  • Cas Number : 141046-52-8
  • Molecular Weight : 184.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1r-4r-4-methoxycarbonyl-bicyclo-2-1-1-hexane-1-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43721
Availability details for
Quantity
  • Availability
  • Add to Cart