ADVH14DBD242 AChemBlock

2,6-dichloropyridine-3-sulfonyl chloride

  • MDL number MFCD06739118
  • Cas Number : 239810-43-6
  • Molecular Weight : 246.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-6-dichloropyridine-3-sulfonyl-chloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43323
Availability details for
Quantity
  • Availability
  • Add to Cart