ADVH14DBD2B9 AChemBlock

4-(3-(Trifluoromethyl)-3H-diazirin-3-yl)-2-hydroxybenzalde

  • MDL number MFCD30468740
  • Cas Number : 308085-25-8
  • Molecular Weight : 230.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-3-trifluoromethyl-3h-diazirin-3-yl-2-hydroxybenzalde-hyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43304
Availability details for
Quantity
  • Availability
  • Add to Cart