ADVH14DBD36D AChemBlock

4-bromo-3-methyl-1H-pyrrolo[2,3-c]pyridine

  • MDL number MFCD31705092
  • Cas Number : 1639444-93-1
  • Molecular Weight : 211.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-3-methyl-1h-pyrrolo-2-3-c-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P47435
Availability details for
Quantity
  • Availability
  • Add to Cart