ADVH14DBD3A9 AChemBlock

5-chloro-1H-pyrrolo[2,3-c]pyridine-2-carboxylic acid

  • MDL number MFCD09878688
  • Cas Number : 800401-68-7
  • Molecular Weight : 196.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-1h-pyrrolo-2-3-c-pyridine-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P44458
Availability details for
Quantity
  • Availability
  • Add to Cart