ADVH14DBD3B5 AChemBlock

5-chloro-1H-pyrrolo[3,2-b]pyridine-2-carboxylic acid

  • MDL number MFCD18261123
  • Cas Number : 800401-63-2
  • Molecular Weight : 196.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-1h-pyrrolo-3-2-b-pyridine-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P44118
Availability details for
Quantity
  • Availability
  • Add to Cart