ADVH14DBD41C AChemBlock

4-bromo-5-methoxy-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD12963485
  • Cas Number : 1190321-71-1
  • Molecular Weight : 227.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-5-methoxy-1h-pyrrolo-2-3-b-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P49474
Availability details for
Quantity
  • Availability
  • Add to Cart