ADVH14DBD45A AChemBlock

trans-3-amino-2,2-dimethylcyclobutanol hydrochloride

  • MDL number MFCD27956965
  • Cas Number : 1434142-03-6
  • Molecular Weight : 151.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/trans-3-amino-2-2-dimethylcyclobutanol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P48170
Availability details for
Quantity
  • Availability
  • Add to Cart