ADVH14DBD498 AChemBlock

8-oxabicyclo[3.2.1]octan-3-ol

  • MDL number MFCD09866852
  • Cas Number : 1172846-34-2
  • Molecular Weight : 128.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/8-oxabicyclo-3-2-1-octan-3-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P49665
Availability details for
Quantity
  • Availability
  • Add to Cart