ADVH14DBD4EC AChemBlock

2-cyclobutylacetic acid

  • MDL number MFCD06411067
  • Cas Number : 6540-33-6
  • Molecular Weight : 114.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-cyclobutylacetic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P44572
Availability details for
Quantity
  • Availability
  • Add to Cart