ADVH14DBD536 AChemBlock

(1S,2S)-2-{[(tert-butoxy)carbonyl]amino}cyclohexane-1-carboxylic acid

  • MDL number MFCD04974226
  • Cas Number : 488703-60-2
  • Molecular Weight : 243.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1s-2s-2-tert-butoxy-carbonyl-amino-cyclohexane-1-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P45567
Availability details for
Quantity
  • Availability
  • Add to Cart