ADVH14DBD5C4 AChemBlock

ethyl 3-oxobicyclo[3.2.1]octane-8-carboxylate

  • MDL number MFCD29921483
  • Cas Number : 172288-92-5
  • Molecular Weight : 196.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-3-oxobicyclo-3-2-1-octane-8-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P47297
Availability details for
Quantity
  • Availability
  • Add to Cart