ADVH14DBD62E AChemBlock

methyl 5-(trifluoromethoxy)-1H-indazole-3-carboxylate

  • MDL number MFCD12498703
  • Cas Number : 932041-12-8
  • Molecular Weight : 260.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-trifluoromethoxy-1h-indazole-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43175
Availability details for
Quantity
  • Availability
  • Add to Cart