ADVH14DBD678 AChemBlock

[cis-4-methylcyclohexyl]methanol

  • MDL number MFCD31705071
  • Cas Number : 3937-48-2
  • Molecular Weight : 128.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/cis-4-methylcyclohexyl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P45661
Availability details for
Quantity
  • Availability
  • Add to Cart