ADVH14DBD798 AChemBlock

4-fluoro-1H-pyrrolo[3,2-c]pyridine-2-carbaldehyde

  • MDL number MFCD31705150
  • Cas Number : 2231674-89-6
  • Molecular Weight : 164.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-fluoro-1h-pyrrolo-3-2-c-pyridine-2-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P50413
Availability details for
Quantity
  • Availability
  • Add to Cart