ADVH14DBD7BB AChemBlock

rel-(1r,4r)-4-amino-1-methylcyclohexane-1-carbonitrile

  • MDL number MFCD31705236
  • Cas Number : 1455039-14-1
  • Molecular Weight : 138.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/rel-1r-4r-4-amino-1-methylcyclohexane-1-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P48082
Availability details for
Quantity
  • Availability
  • Add to Cart