ADVH14DBD841 AChemBlock

1-methyl-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD09743451
  • Cas Number : 27257-15-4
  • Molecular Weight : 132.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-methyl-1h-pyrrolo-2-3-b-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P44494
Availability details for
Quantity
  • Availability
  • Add to Cart