ADVH14DBD8B3 AChemBlock

4-(propan-2-yloxy)cyclohexan-1-one

  • MDL number MFCD16658578
  • Cas Number : 69697-46-7
  • Molecular Weight : 156.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-propan-2-yloxy-cyclohexan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P45305
Availability details for
Quantity
  • Availability
  • Add to Cart