ADVH14DBD923 AChemBlock

1-methoxycarbonylcyclopropanecarboxylic acid

  • MDL number MFCD06800496
  • Cas Number : 113020-21-6
  • Molecular Weight : 144.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-methoxycarbonylcyclopropanecarboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P49754
Availability details for
Quantity
  • Availability
  • Add to Cart