ADVH14DBD937 AChemBlock

trans-4-ethynylcyclohexan-1-amine hydrochloride

  • MDL number MFCD29046545
  • Cas Number : 2095396-33-9
  • Molecular Weight : 159.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/trans-4-ethynylcyclohexan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P46268
Availability details for
Quantity
  • Availability
  • Add to Cart