ADVH14DBD96F AChemBlock

2,2-difluorocyclohexan-1-ol

  • MDL number MFCD28635565
  • Cas Number : 1546282-70-5
  • Molecular Weight : 136.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-2-difluorocyclohexan-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P47901
Availability details for
Quantity
  • Availability
  • Add to Cart