ADVH14DBD9E1 AChemBlock

methyl 3-fluorocyclobutane-1-carboxylate

  • MDL number MFCD20482254
  • Cas Number : 1547065-03-1
  • Molecular Weight : 132.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-fluorocyclobutane-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P47896
Availability details for
Quantity
  • Availability
  • Add to Cart