ADVH14DBD9E8 AChemBlock

methyl[(1-methylazetidin-3-yl)methyl]amine

  • MDL number MFCD28046336
  • Cas Number : 864350-83-4
  • Molecular Weight : 114.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-methylazetidin-3-yl-methyl-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P44272
Availability details for
Quantity
  • Availability
  • Add to Cart