ADVH14DBDA00 AChemBlock

N-[1-(diphenylmethyl)azetidin-3-yl]acetamide

  • MDL number MFCD06808718
  • Cas Number : 102065-87-2
  • Molecular Weight : 280.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/n-1-diphenylmethyl-azetidin-3-yl-acetamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P49964
Availability details for
Quantity
  • Availability
  • Add to Cart