ADVH14DBDA0A AChemBlock

N-(2-methoxyethyl)-N-methylazetidin-3-amine

  • MDL number MFCD11849036
  • Cas Number : 759436-38-9
  • Molecular Weight : 144.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/n-2-methoxyethyl-n-methylazetidin-3-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P45215
Availability details for
Quantity
  • Availability
  • Add to Cart