ADVH14DBDA1D AChemBlock

(azetidin-2-yl)methanamine hydrochloride

  • MDL number MFCD09997790
  • Cas Number : 103550-76-1
  • Molecular Weight : 122.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/azetidin-2-yl-methanamine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P49927
Availability details for
Quantity
  • Availability
  • Add to Cart