ADVH14DBDA24 AChemBlock

tert-butyl 3-ethyl-3-hydroxyazetidine-1-carboxylate

  • MDL number MFCD09264470
  • Cas Number : 398489-28-6
  • Molecular Weight : 201.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-ethyl-3-hydroxyazetidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P45658
Availability details for
Quantity
  • Availability
  • Add to Cart