ADVH14DBDA26 AChemBlock

N,3-dimethylazetidin-3-amine

  • MDL number MFCD09264399
  • Cas Number : 149696-11-7
  • Molecular Weight : 100.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/n-3-dimethylazetidin-3-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P48000
Availability details for
Quantity
  • Availability
  • Add to Cart