ADVH14DBDA30 AChemBlock

trans-N1,N1-dimethylcyclobutane-1,3-diamine

  • MDL number MFCD22396942
  • Cas Number : 1821832-50-1
  • Molecular Weight : 114.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/trans-n1-n1-dimethylcyclobutane-1-3-diamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P46926
Availability details for
Quantity
  • Availability
  • Add to Cart