ADVH14DBDA3D AChemBlock

5,8-dioxaspiro[3.4]octan-2-one

  • MDL number MFCD24712320
  • Cas Number : 1824626-99-4
  • Molecular Weight : 128.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-8-dioxaspiro-3-4-octan-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P46871
Availability details for
Quantity
  • Availability
  • Add to Cart