ADVH14DBDA5A AChemBlock

4-methoxy-4-methylcyclohexane-1-carbaldehyde

  • MDL number MFCD28501639
  • Cas Number : 1637310-67-8
  • Molecular Weight : 156.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-methoxy-4-methylcyclohexane-1-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P43770
Availability details for
Quantity
  • Availability
  • Add to Cart