ADVH14DBDA77 AChemBlock

{2-oxabicyclo[2.2.2]octan-1-yl}methanol

  • MDL number MFCD28361974
  • Cas Number : 1447943-03-4
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-oxabicyclo-2-2-2-octan-1-yl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P48109
Availability details for
Quantity
  • Availability
  • Add to Cart